missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Phenyl Sepharose™ 6 Fast Flow Chromatography Media

Used for capture and intermediate purification requiring low to medium hydrophobicity

Brand:  Cytiva 17-0973-05


 View more versions of this product

Product Code. 10343340

  • £630.00 / 200mL
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Base matrix derivatized via uncharged, chemically-stable ether linkages
  • Two levels of substitution help find optimal selectivity and binding capacity for given application
  • Hydrophilic nature of the base matrix ensures low levels of non-specific binding
  • Hydrophobic interaction chromatography (HIC) media composed of cross-linked 6% agarose beads modified with standard aromatic phenyl groups via uncharged, chemically-stable ether linkages
  • Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development
  • Degree of substitution for low sub HIC media is ∽25μmol phenyl/mL gel
  • Degree of substitution for high sub HIC media is ∽40μmol phenyl/mL gel

TRUSTED_SUSTAINABILITY
Specifications

Specifications

200 mL
For Capture and Intermediate PurifiCatIon Requiring Low to Medium Hydrophobicity
Phenyl Sepharose 6 Fast Flow Media (high sub)
45 to 165 μm
3 to 13
Phenyl
Stable in commonly used aqueous Buffers: 1 M NaOH, 3 M Ammonium Sulphate, 30% Isopropanol, 70% Ethanol, 10% Ethylene Glycol, 0.5% SDS, 6 M Guanidine-Hydrochloride, 8 M Urea
4°C to 30°C
250 to 400 cm/Hoμr
6% Cross-Linked Agarose
20% Ethanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Phenyl Sepharose 6 Fast Flow (high sub), 200 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.