Learn More
Tocris Bioscience™ Phortress
Prodrug of the antitumor agent 5F 203
Brand:  Tocris Bioscience™ 4995/10
115.76 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18759007
  • £145.00 / 10mg
Explore more special offers
Description
Description
Phortress is a prodrug of the antitumor agent 5F 203, which acts via binding to aryl hydrocarbon receptors. Induces expression of CYP1A1 and generates adducts in the DNA of sensitive MCF7 and IGROV-1 cells.
Specifications
Specifications
>98%
Phortress
Phortress
10 mg
459.41