missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Phortress
Prodrug of the antitumor agent 5F 203
£145.00 - £526.00
Specifications
Purity | >98% |
---|---|
Product Type | Phortress |
Molecular Weight (g/mol) | 459.41 |
Inhibitors | Phortress |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18759007
|
Tocris Bioscience™
4995/10 |
10 mg |
£145.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18799007
|
Tocris Bioscience™
4995/50 |
50 mg |
£526.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Prodrug of the antitumor agent 5F 203, which acts via binding to aryl hydrocarbon receptors. Induces expression of CYP1A1 and generates adducts in the DNA of sensitive MCF7 and IGROV-1 cells.Specifications
>98% | |
Phortress | |
459.41 | |
Phortress |
For Research Use Only