Learn More
Tocris Bioscience™ Phortress
Prodrug of the antitumor agent 5F 203
Brand: Tocris Bioscience™ 4995/50
420.72 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Phortress is a prodrug of the antitumor agent 5F 203, which acts via binding to aryl hydrocarbon receptors. Induces expression of CYP1A1 and generates adducts in the DNA of sensitive MCF7 and IGROV-1 cells.Specifications
>98% | |
Phortress | |
Phortress |
50 mg | |
459.41 |
For Research Use Only