missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phosphatase Inhibitor Cocktail I

 

Brand:  Thermo Scientific Chemicals J63907.AA

105.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.



Product Code. 15416069

  • £105.53 / 5mL
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Phosphatase Inhibitor Cocktail I is used to inhibitors with a broad specificity for serine, cysteine, and acid proteases, and aminopeptidases. It has been tested on a wide variety of cells and tissue extracts to isolate/maintain intact proteins without degradation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Colorless
-30°C to -10°C
5 mL
Phosphatase Inhibitor Cocktail
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Phosphatase inhibitors cocktail I

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H360D-May damage the unborn child.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Trisodium orthovanadate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only