Learn More
Thermo Scientific Chemicals Phosphate-buffered saline (PBS, 10X), pH 7.4, for Western blot
For Western blot washing
Brand: Thermo Scientific Chemicals J60801.K7
149.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Phosphate-buffered saline (PBS, 10X), is an isotonic, non-toxic to most cells and is used in biological research. It is associated with ethylenediaminetetraacetic acid (EDTA) and used to disengage attached and clumped cells. It also helps to maintain a constant pH.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Colorless | |
Phosphate Buffered Saline (PBS) | |
For Western blot washing. | |
Liquid |
10 X | |
4 L | |
7.4 |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Z27K524 | Certificate of Analysis | 23/01/2025 | J60801.K7,J60801.K3,J60801.K2 |
Z27K523 | Certificate of Analysis | 21/01/2025 | J60801.K7,J60801.K3,J60801.K2 |
Y18K553 | Certificate of Analysis | 06/12/2024 | J60801.K7,J60801.K3,J60801.K2 |
U25K530 | Certificate of Analysis | 11/09/2024 | J60801.K7,J60801.K3,J60801.K2 |
S01K545 | Certificate of Analysis | 26/07/2024 | J60801.K7,J60801.K3,J60801.K2 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only