Learn More
Indole-3-butyric acid, MedChemExpress
MedChemExpress Indole-3-butyric acid (3-indolebutyric acid; IBA) is a plant growth auxin and a good rooting agent. It can promote herbs and woody ornamental plant rooting and used for improving fruit rate.
Brand: Medchem Express HY-N0186

Description
Indole-3-butyric acid (3-indolebutyric acidSpecifications
Indole-3-butyric acid | |
Off-White | |
C12H13NO2 | |
3-indolebutyric acid | |
O=C(O)CCCC1=CNC2=C1C=CC=C2 | |
203.24 | |
Research | |
Metabolism-sugar/lipid metabolism |
133-32-4 | |
Research | |
10 g | |
DMSO : ≥ 35 mg/mL (172.21 mM) | |
203.24 | |
99.9% | |
Solid |
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only