Learn More
Phosphorus tribromide, 1.0M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 382061000
40.40 GBP valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
Specifications
Phosphorus tribromide | |
7789-60-8, 75-09-2 | |
Liquid | |
Br3P | |
MFCD00011436 | |
phosphorus tribromide, phosphorous tribromide, tribromophosphine, phosphorus iii bromide, phosphorous bromide, pbr3, phosphorus bromide, phosphorus bromide pbr3, unii-58r3866pua, tribro-mophosphine | |
P(Br)(Br)Br | |
270.69 | |
270.69 | |
AcroSeal™ Glass bottle | |
1.4880g/mL |
1.0M Solution in Dichloromethane | |
Colorless to Yellow | |
100 mL | |
PBr2 | |
15, 7469 | |
IPNPIHIZVLFAFP-UHFFFAOYSA-N | |
tribromophosphane | |
24614 | |
0.9 to 1.1M | |
1.488 |
Safety and Handling
- Phosphorus tribromide, 1M solution in dichloromethane
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H351-Suspected of causing cancer.
- H373-May cause damage to organs.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Special Offers
-
Multiple Manufacturers
New Offer Save Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only