missing translation for 'onlineSavingsMsg'
Learn More
Phosphorus(V) oxybromide, 98% min, Thermo Scientific Chemicals Product Code.: 11498660

Phosphorus(V) oxybromide, 98% min, Thermo Scientific Chemicals

Product Code. 11498660
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11498660

Brand: Thermo Scientific Alfa Aesar 056135.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use As a Chemical Intermediate

As a chemical intermediate.Phosphorus(V) oxybromide is used as a brominating agent in the synthesis of 2,4,6-tribromopyrimidine and bromoheterocyclic compounds. It is used in the phosphorus flame retardant and phosphorus activators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-59-5
Molecular Formula Br3OP
Molecular Weight (g/mol) 286.685
MDL Number MFCD00036298
InChI Key UXCDUFKZSUBXGM-UHFFFAOYSA-N
Synonym phosphorus oxybromide, phosphoric tribromide, phosphoryl bromide, phosphoryl tribromide, phosphorusoxybromide, phosphorus v oxybromide, unii-h98h8y87qq, phosphorousoxybromide, phosphoroyl tribromide, pobr3Show MoreShow Less
PubChem CID 24613
SMILES O=P(Br)(Br)Br

Specifications

Boiling Point 189°C
Melting Point 56°C
Physical Form Lump
Quantity 100 g
UN Number UN1939
Merck Index 14,7348
Solubility Information Decomposes in water; Soluble in ether,carbon disulfide,benzene,chloroform and sulfuric acid.
Formula Weight 286.73
Percent Purity ≥98%
Sensitivity Moisture sensitive
Density 2.822
Chemical Name or Material Phosphorus(V) oxybromide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS27K039 Certificate TypeCertificate of Analysis Date09/07/2024 Catalog Number056135.22,056135.14,056135.06
Lot NumberQ16K035 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number056135.22,056135.14,056135.06
Lot NumberN17J039 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number056135.22,056135.14,056135.06
Lot NumberP31I039 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog Number056135.22,056135.14,056135.06
Lot NumberI15R005 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number056135.22,056135.14,056135.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phosphorus(V) oxybromide, 98% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.