missing translation for 'onlineSavingsMsg'
Learn More

Palintest™ Photometer Kit

This unique photometer kit comes complete in a case with six glass tubes and one dilution tube for the photometer 7100 standard model device.

Brand:  Palintest™ PTH7100

 View more versions of this product

Product Code. 10676325

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

  • Includes: 7100 Model Standard, with accessories
  • Photometer kit for 7100 device
  • Comes complete in case with six glass tubes and one dilution tube 
  • Power supply 
  • 3 AA batteries
  • L x W x H: 146 x 275 x 75 mm
  • Weight: 975 g
     
Specifications

Specifications

Kit for Photometer 7100 device for field measurements
  • Includes: 7100 Model Standard, with accessories
  • Complete in case with six glass tubes and one dilution tube
  • Power supply
  • 3 AA batteries 
  • L x W x H: 146 x 275 x 75 mm.
  • Weight: 975 g
  • Complete in case with 6 glass tubes and a tube for dilution
Photometer Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Palintest™ Photometer Kit > Includes: 7100 Model Standard, with accessories

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.