Learn More
Thermo Scientific™ Phusion™ Hot Start II DNA Polymerase (2 U/μL)
Thermo Scientific Phusion High-Fidelity DNA Polymerases set a gold standard for high performance PCR.
Brand: Thermo Scientific™ F549XL
1596.98 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : CAS Number : 9002-93-1 Weight : 0.19200kg
Description
Thermo Scientific Phusion High-Fidelity DNA Polymerases set a gold standard for high performance PCR. Featuring an error rate 50-fold lower than that of Taq, and 6-fold lower than that of Pfu, Phusion High-Fidelity DNA Polymerase is excellent choice for cloning and other applications requiring high fidelity. Phusion DNA Polymerases offer robust performance with short protocol times, even in the presence of PCR inhibitors, and generate higher yields with lower enzyme amounts than other DNA polymerase.
Using Phusion Hot Start II High-Fidelity DNA Polymerase, amplification proceeds without the production of nonspecific products due to the combination of Phusion DNA Polymerase and a reversibly bound, specific Affibody ligand that inhibits DNA polymerase activity at room temperature.
The Affibody ligand also inhibits the 3´→5´ exonuclease activity of the polymerase, thus preventing degradation of primers and template DNA during reaction set up. At temperatures that promote polymerase activity, the ligand is released, rendering the polymerase fully active. Phusion Hot Start II DNA Polymerase is immediately reactivated at high temperatures so it does not require a separate activation step in PCR protocols.
Highlights
• Reaction set up at room temperature
• No non-specific amplification and primer degradation during reaction set up
• Zero-time reactivation due to unique hot start technology
• High fidelity (52X Taq)
• Fast PCR due to short extension times (15-30 s/kb)
• Robust reactions, minimal optimization needed
• Increased product yields with minimal enzyme amounts
• Available as a Green buffer format permitting direct loading on gels (F-537S or F-537L)
Applications
• High-fidelity PCR
• High throughput
• Amplification of difficult (GC-rich) templates
• Template generation for sequencing
• Multiplex PCR
• Long-range PCR
• Cloning
• Mutagenesis
• Microarray
Using Phusion DNA Polymerases
Annealing rules for Phusion DNA Polymerases are different from many common DNA polymerases (such as Taq DNA polymerases). For optimal results, use our Tm calculator at www.thermofisher.com/tmcalculator.
Specifications
• Phusion Hot Start II High-Fidelity DNA Polymerase (2 U/μL) • 5X Phusion HF & GC Buffers • DMSO • 50 mM MgCl2 solution Phusion HF Buffer and Phusion GC Buffers each provide 1.5 mM MgCl2 in the final 1X concentration. Store at -20°C. |
|
Phusion Hot Start II DNA Polymerase | |
Hot Start DNA Polymerase and dNTP Mix | |
Hot-start PCR, High-fidelity PCR | |
52X | |
2000 Reactions | |
Standalone |
High | |
Fast | |
Dry Ice | |
2 U/μL | |
Built-In Hot Start | |
Blunt | |
20 kb or less |
For Research Use Only. Not for use in diagnostic procedures.