missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phusion U Hot Start DNA Polymerases

Thermo Scientific Phusion U DNA polymerase is a novel engineered high fidelity enzyme developed using fusion technology.

Brand:  Thermo Scientific™ F555L

458.90 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 15230143

  • £706.00 / 500 units

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific Phusion U DNA Polymerase is a novel engineered high fidelity enzyme developed using fusion technology. Due to a proprietary mutation in the so called dUTP binding pocket of Phusion polymerase, Phusion U polymerase overcomes an important limitation of proofreading enzymes: it is able to incorporate dUTP and read through uracil present in DNA templates.

In addition to the uracil usage possibility, Phusion U polymerase features all the superior properties of other Phusion DNA polymerases: great accuracy, speed, ability to amplify long amplicons up to 20 kb, and a high specificity with Affibody-based hot start. These features make Phusion U Hot Start DNA Polymerase an excellent choice for such important applications as amplification of bisulphite-converted or damaged DNA as well as use of carryover contamination control.

Phusion U Hot Start Green DNA Polymerase is a combination of Phusion U Hot Start DNA Polymerase and 5X Phusion Green buffers. The buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with Phusion U polymerase performance and is compatible with downstream applications such as DNA sequencing, ligation, and restriction digestion.

Highlights

  • Accuracy—high fidelity DNA polymerase (25X Taq)
  • Uracil-tolerance—engineered to incorporate dUTP and amplify uracil containing templates
  • Specificity—hot start for reduced nonspecific amplification and primer degradation
  • Speed—short extension times (15–30 s/kb)
  • Green format—permits direct loading of PCR products on gels

Applications

  • Amplification of bisulphite-converted DNA
  • Amplification of damaged or aged DNA
  • Carry-over contamination control
  • Uracil-excision based (USER) cloning methods

Using Phusion DNA polymerases

Annealing rules for Phusion DNA Polymerases are different from many common DNA polymerases (such as Taq DNA polymerases).

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• Phusion U Hot Start DNA Polymerase, 2 U/μL
• 5X Phusion HF & GC Buffers
• DMSO
• 50 mM MgCl2 solution

Phusion HF Buffer and Phusion GC Buffers each provide 1.5 mM MgCl2 in the final 1X concentration.
Store at -20°C.
Phusion U Hot Start DNA Polymerase
Hot Start DNA Polymerase
Dry Ice
2 U/μL
Built-In Hot Start
Blunt
20 kb or less
High
Fast
500 units
Hot-start PCR, High-fidelity PCR
25X
500 Reactions
Standalone
Colorless
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.