Learn More
Thermo Scientific™ Phusion U Hot Start DNA Polymerases
Thermo Scientific Phusion U DNA polymerase is a novel engineered high fidelity enzyme developed using fusion technology.
£159.00 - £716.00
Specifications
GC-Rich PCR Performance | High |
---|---|
Polymerase | Phusion U Hot Start DNA Polymerase |
Reaction Speed | Fast |
Product Type | Hot Start DNA Polymerase |
Shipping Condition | Dry Ice |
Product Code | Brand | Color | No. of Reactions | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | No. of Reactions | Price | Quantity & Availability | ||||
15250143
|
Thermo Scientific™
F556L |
Green | 500 Reactions |
£716.00
500 units |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15240143
|
Thermo Scientific™
F556S |
Green | 100 Reactions |
£159.00
100 units |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15230143
|
Thermo Scientific™
F555L |
Colorless | 500 Reactions |
£706.00
500 units |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15220143
|
Thermo Scientific™
F555S |
Colorless | 100 Reactions |
£159.00
100 units |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Thermo Scientific Phusion U DNA Polymerase is a novel engineered high fidelity enzyme developed using fusion technology. Due to a proprietary mutation in the so called dUTP binding pocket of Phusion polymerase, Phusion U polymerase overcomes an important limitation of proofreading enzymes: it is able to incorporate dUTP and read through uracil present in DNA templates.
In addition to the uracil usage possibility, Phusion U polymerase features all the superior properties of other Phusion DNA polymerases: great accuracy, speed, ability to amplify long amplicons up to 20 kb, and a high specificity with Affibody-based hot start. These features make Phusion U Hot Start DNA Polymerase an excellent choice for such important applications as amplification of bisulphite-converted or damaged DNA as well as use of carryover contamination control.
Phusion U Hot Start Green DNA Polymerase is a combination of Phusion U Hot Start DNA Polymerase and 5X Phusion Green buffers. The buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with Phusion U polymerase performance and is compatible with downstream applications such as DNA sequencing, ligation, and restriction digestion.
Highlights
- Accuracy—high fidelity DNA polymerase (25X Taq)
- Uracil-tolerance—engineered to incorporate dUTP and amplify uracil containing templates
- Specificity—hot start for reduced nonspecific amplification and primer degradation
- Speed—short extension times (15–30 s/kb)
- Green format—permits direct loading of PCR products on gels
Applications
- Amplification of bisulphite-converted DNA
- Amplification of damaged or aged DNA
- Carry-over contamination control
- Uracil-excision based (USER) cloning methods
Using Phusion DNA polymerases
Annealing rules for Phusion DNA Polymerases are different from many common DNA polymerases (such as Taq DNA polymerases).
Specifications
High | |
Fast | |
Dry Ice | |
2 U/μL | |
Built-In Hot Start | |
Standalone |
Phusion U Hot Start DNA Polymerase | |
Hot Start DNA Polymerase | |
Hot-start PCR, High-fidelity PCR | |
25X | |
Blunt | |
20 kb or less |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.