missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PI 828, Tocris Bioscience™
PI 3-kinase inhibitor, more potent than LY 294002 (Cat. No. 1130)
£117.31
Chemical Identifiers
CAS | 942289-87-4 |
---|---|
Molecular Formula | C19H18N2O3 |
Molecular Weight (g/mol) | 322.364 |
InChI Key | WUKMIBOGGXMBAC-UHFFFAOYSA-N |
Synonym | 8-4-aminophenyl-2-morpholino-4h-chromen-4-one, ly 294002, 4gamma-nh2, 2-4-morpholinyl-8-4-aminopheny l-4h-1-benzopyran-4-one, 8-4-aminophenyl-2-4-morpholinyl-4h-1-benzopyran-4-one, 8-4-aminophenyl-2-morpholin-4-yl-4h-1-benzopyran-4-one, 8-4-aminophenyl-2-morpholin-4-yl chromen-4-one |
PubChem CID | 25181195 |
IUPAC Name | 8-(4-aminophenyl)-2-morpholin-4-ylchromen-4-one |
SMILES | C1COCCN1C2=CC(=O)C3=C(O2)C(=CC=C3)C4=CC=C(C=C4)N |
Description
PI 3-Kinase inhibitor (IC50 values are 0.098, 0.183, 0.227 and 1.967 μM for p110β, p110α, p110δ and p110γ respectively) that displays higher potency than LY 294002 (Cat No. 1130). Can be immobilized onto a solid phase.Chemical Identifiers
942289-87-4 | |
322.364 | |
8-4-aminophenyl-2-morpholino-4h-chromen-4-one, ly 294002, 4gamma-nh2, 2-4-morpholinyl-8-4-aminopheny l-4h-1-benzopyran-4-one, 8-4-aminophenyl-2-4-morpholinyl-4h-1-benzopyran-4-one, 8-4-aminophenyl-2-morpholin-4-yl-4h-1-benzopyran-4-one, 8-4-aminophenyl-2-morpholin-4-yl chromen-4-one | |
8-(4-aminophenyl)-2-morpholin-4-ylchromen-4-one |
C19H18N2O3 | |
WUKMIBOGGXMBAC-UHFFFAOYSA-N | |
25181195 | |
C1COCCN1C2=CC(=O)C3=C(O2)C(=CC=C3)C4=CC=C(C=C4)N |
Specifications
942289-87-4 | |
8-4-aminophenyl-2-morpholino-4h-chromen-4-one, ly 294002, 4gamma-nh2, 2-4-morpholinyl-8-4-aminopheny l-4h-1-benzopyran-4-one, 8-4-aminophenyl-2-4-morpholinyl-4h-1-benzopyran-4-one, 8-4-aminophenyl-2-morpholin-4-yl-4h-1-benzopyran-4-one, 8-4-aminophenyl-2-morpholin-4-yl chromen-4-one | |
C1COCCN1C2=CC(=O)C3=C(O2)C(=CC=C3)C4=CC=C(C=C4)N | |
322.364 | |
322.36 | |
PI 828 |
C19H18N2O3 | |
WUKMIBOGGXMBAC-UHFFFAOYSA-N | |
8-(4-aminophenyl)-2-morpholin-4-ylchromen-4-one | |
25181195 | |
>99% |
Safety and Handling
Recommended Storage : Store at +4°C