Learn More
Thermo Scientific™ Pierce™ Agarose ChIP Kit
Chromatin immunoprecipitation (chromatin IP, ChIP) assay kit containing all reagents, agarose affinity beads, and a simple, fast, reliable protocol.
Brand: Thermo Scientific™ 26156
Code : PN
Additional Details : Weight : 0.10000kg Transport : UN number : 1219 Chem class : 3 Pack group : II
Description
Thermo Scientific™ Pierce Agarose ChIP Kit provides a complete set of reagents and a simple, fast and reproducible protocol to perform chromatin immunoprecipitation (ChIP) assays.
This chromatin IP kit contains sufficient reagents to perform 30 ChIP assays with appropriate controls using an optimized protocol and a convenient spin-column format. The kit includes a complete Chromatin Prep Module (also available separately) to efficiently extract and solubilize crosslinked DNA-protein interaction complexes from cells, plus an immunoprecipitation module that contains high-capacity, ChIP-grade Protein A/G agarose.Highlights:
- Simple and streamlined ChIP assay protocol (< 8 hours)
- Highly efficient isolation and lysis of nuclei
- Easy and reproducible enzymatic digestion
- Low-background and high-capacity Protein A/G agarose resin
- Highly specific positive control reagents included: RNA polymerase II antibody and GAPDH PCR primers
- Fast and reproducible spin-column format; columns and collection tubes included
- High-recovery DNA purification
- Choose a complete ChIP assay kit or the separate chromatin preparation module
Includes:
Kit contains the Chromatin Prep Module (Part No. 26158; 11 components) for chromatin extraction and a complete set of immunoprecipitation reagents (15 components) for IP experiments with controls
Specifications
>50 mg Human IgG/mL resin binding capacity | |
Pierce Agarose ChIP Kit | |
Pierce™ | |
ChIP Kit | |
Kit |
≥50 mg Human IgG per mL Resin Binding | |
Sufficient For: 30 complete chromatin IP assay experiments• ChIP Grade Protein A/G Plus Agarose, 0.65 mL (store at 4°C) • IP Dilution/Wash Buffer (5X), 11 mL (store at 4°C) • IP Wash Buffer 3 (5X), 4.5 mL (store at 4°C) • IP Elution Buffer (2X), 4.5 mL (store at 4°C) • Column Accessory Pack, 3 packs (store at room temperature or 4°C) • Microcentrifuge Tubes (1.5mL), 75 tubes (store at room temperature or 4°C) • DNA Clean-Up Column, 40 columns (store at room temperature or 4°C) • DNA Column Binding Solution, 30 mL (store at room temperature or 4°C) • DNA Column Wash Solution, 6 mL (store at room temperature or 4°C) • pH Indicator, 0.8 mL (store at room temperature or 4°C) • DNA Column Elution Solution, 5 mL (store at room temperature or 4°C) • Anti-RNA Polymerase II Antibody, 25 μL (store at -20°C) • Normal Rabbit IgG (1 mg/mL), 10 μL |
|
30 Reaction Kit | |
30 Reactions |
For Research Use Only. Not for use in diagnostic procedures.