missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Alkaline Phosphatase

Purified calf-intestinal alkaline phosphatase (AP, alk-phos) enzyme for use in activity assays and conjugation to antibodies for ELISA and IHC.

Brand:  Thermo Scientific™ 31391

 View more versions of this product

Product Code. 10076073

  • £960.00 / 20mg
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Alkaline Phosphatase is purified calf-intestinal alkaline phosphatase (AP or alk-phos) enzyme for use in activity assays and conjugation to antibodies for ELISA and immunohistochemistry applications.

Highlights:

  • Purified form - ready to conjugate without prior dialysis
  • Activity is not affected by exposure to antibacterial agents, such as sodium azide or thimerosal
  • Specific activity > 2,000 units/mg
  • One unit is defined as the amount that will hydrolyze 1.0 mol of p-nitrophenyl phosphate per minute at 37° C in 1.0M diethanolamine, 0.5mM MgCl2, pH 7.8
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at 2–8°C.
ELISA, Immunohistochemistry
AP (Alkaline Phosphatase)
Solid
Pierce™
20 mg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.