missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ Alkaline Phosphatase
Purified calf-intestinal alkaline phosphatase (AP, alk-phos) enzyme for use in activity assays and conjugation to antibodies for ELISA and IHC.
£960.00 - £3755.00
Specifications
Content And Storage | Store at 2–8°C. |
---|---|
For Use With (Application) | ELISA, Immunohistochemistry |
Product Line | Pierce™ |
Label or Dye | AP (Alkaline Phosphatase) |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10076073
|
Thermo Scientific™
31391 |
20mg |
£960.00
20mg |
In Stock
Log in to see stock. |
|||||
10299803
|
Thermo Scientific™
31392 |
5 x 20mg |
£3755.00
100mg |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
Description
Thermo Scientific™ Pierce Alkaline Phosphatase is purified calf-intestinal alkaline phosphatase (AP or alk-phos) enzyme for use in activity assays and conjugation to antibodies for ELISA and immunohistochemistry applications.
Highlights:
- Purified form - ready to conjugate without prior dialysis
- Activity is not affected by exposure to antibacterial agents, such as sodium azide or thimerosal
- Specific activity > 2,000 units/mg
- One unit is defined as the amount that will hydrolyze 1.0 mol of p-nitrophenyl phosphate per minute at 37° C in 1.0M diethanolamine, 0.5mM MgCl2, pH 7.8
Specifications
Store at 2–8°C. | |
ELISA, Immunohistochemistry | |
Pierce™ | |
AP (Alkaline Phosphatase) |
For Research Use Only. Not for use in diagnostic procedures.