Learn More
Thermo Scientific™ Pierce™ Alkaline Phosphatase
Purified calf-intestinal alkaline phosphatase (AP, alk-phos) enzyme for use in activity assays and conjugation to antibodies for ELISA and IHC.
Brand: Thermo Scientific™ 31392
Code : PN
Additional Details : Weight : 0.00010kg
Description
Thermo Scientific™ Pierce Alkaline Phosphatase is purified calf-intestinal alkaline phosphatase (AP or alk-phos) enzyme for use in activity assays and conjugation to antibodies for ELISA and immunohistochemistry applications.
Highlights:
- Purified form - ready to conjugate without prior dialysis
- Activity is not affected by exposure to antibacterial agents, such as sodium azide or thimerosal
- Specific activity > 2,000 units/mg
- One unit is defined as the amount that will hydrolyze 1.0 mol of p-nitrophenyl phosphate per minute at 37° C in 1.0M diethanolamine, 0.5mM MgCl2, pH 7.8
Specifications
Store at 2–8°C. | |
ELISA, Immunohistochemistry | |
AP (Alkaline Phosphatase) |
Liquid | |
Pierce™ | |
5 x 20 mg |
For Research Use Only. Not for use in diagnostic procedures.