Learn More
Thermo Scientific™ Pierce™ Alkylating Reagents
Alkylating reagents provided in single-use, pre-weighed formats to streamline protein preparation workflows.
Brand:  Thermo Scientific™ A39270
 View more versions of this product
Product Code. 16336562
  • £93.00 / Pack of 10
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermo Scientific Pierce Iodoacetamide or Chloroacetamide are provided in a no-weigh format, where small amounts of reagent are pre-weighed and packaged for single use. This single-use format prevents the contamination and loss of reagent reactivity over time associated with repetitive opening and closing of a single vial. The format enables use of a fresh vial of reagent each time, and eliminates the hassle of weighing out small amounts of reagent.
Alkylating reagents iodoacetamide and chloracetamide are sulfhydryl-reactive alkylating reagents used to block reduced cysteine residues for protein characterization and peptide mapping. Alkylation with iodoacetamide or chloracetamide after cysteine reduction results in the covalent addition of a carbamidomethyl group (57.07 Da) and prevents the formation of disulfide bonds. Reducing agents added after alkylation will react with excess iodoacetamide.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Alkylating Reagent
10 x 2 mg
Chloroacetamide
Pierce™