missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ Anti-DYKDDDDK Affinity Resin
Pierce Anti-DYKDDDDK Affinity Resin provides a fast, convenient method for purification and immunoprecipitation of DYKDDDDK-tagged proteins
Brand: Thermo Scientific™ A36804
7457.50 GBP valid until 2024-12-31
Use promo code "21963" to get your promotional price.
This item is not returnable.
View return policy
Description
- The Thermo Scientific Pierce Anti-DYKDDDDK Affinity Resin provides a fast, convenient method for purification and immunoprecipitation (IP) of DYKDDDDK-tagged proteins expressed in in vitro protein expression systems, bacteria, yeast, and mammalian cells. The amino acid sequence DYKDDDDK, commonly known as “Flag”, is recognized by a high-affinity rat monoclonal antibody (clone L5) that is covalently attached to UltraLink Biosupport.
- For protein purification or IP, resin is added to a sample containing DYKDDDDK-tagged protein(s) with the tag on either the N- or the C-terminus. Following capture of DYKDDDDK-tagged proteins, non-specifically bound proteins can be washed away before dissociating bound target protein(s) with elution buffer. The Pierce Anti-DYKDDDDK Affinity Resin is recommended for use in spin purification columns or cartridges for FPLC instruments.
- UltraLink Biosupport is a rigid, hydrophilic, highly crosslinked, copolymeric and porous resin with high coupling capacity. The porosity, rigidity, and durability of the resin also make it suitable for medium-pressure, fast-flow techniques involving large sample volumes.
- Specific—Unique base-beads and highly specific antibody minimizes off-target binding (low non-specific binding)
- High purity —Optimized Bind-Wash-Elute protocol enables high purity
- High Yield —special antibody conjugation method enables high yield
- Rapid—entire purification protocol typically takes less than 40 mins
- Economical—purification protocol allows multiple reuses
- Versatile—can be used in spin-, gravity- or FPLC-based purification and enrichment methods
- Scalable—available in 1, 5, and 50 mL settled resin package sizes to allow for smaller scale immunoprecipitations or larger scale purifications
Specifications
Pierce™ | |
100 mL | |
Liquid | |
Wet Ice | |
Affinity Resin |
Affinity Resin | |
50 mL settled resin. 100 mL supplied as a 50% slurry in PBS containing 0.02% sodium azide. Store at 2–8°C. | |
Anti-DYKDDDDK monoclonal antibodies | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.