missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Anti-DYKDDDDK Magnetic Agarose

Thermo Scientific™ Pierce™ Anti-DYKDDDK Magnetic Agarose Beads provide a fast, convenient method for purification of DYKDDDK-tagged recombinant proteins

£464.00 - £1498.00

Specifications

Matrix Magnetic Agarose
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15875833
View Documents
Thermo Scientific™
A36797
4 mL
£464.00
1mL
Please to purchase this item. Need a web account? Register with us today!
15883521
View Documents
Thermo Scientific™
A36798
20 mL
£1498.00
5mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thermo Scientific Pierce Anti-DYKDDDDK Magnetic Agarose provides a fast, convenient method for purification and immunoprecipitation (IP) of DYKDDDDK-tagged proteins from in vitro protein expression systems, bacteria, yeast, and mammalian cells. The amino acid sequence DYKDDDDK, commonly known as 'FLAG', is recognized by a high-affinity rat monoclonal antibody (clone L5) that is covalently attached to a magnetite-embedded agarose core particle.

For protein purification, the magnetic agarose is added to a sample containing DYKDDDDK-tagged proteins with the tag on either the N- or the C-terminus. Captured proteins are then magnetically separated from the supernatant, and non-specifically bound proteins can be washed away before dissociating bound DYKDDDDK-tagged proteins with elution buffer. The magnetic agarose is removed from the solution using a magnetic stand or an instrument such as the KingFisher Flex Magnetic Particle Processor. Automated instruments are especially useful for higher throughput purifications and screening of purification conditions.

Features include:

  • Specific—unique base beads and highly specific antibody minimizes off-target binding (low non-specific binding)
  • High purity—optimized bind-wash-elute protocol enables high purity
  • High yield—special antibody conjugation method enables high yield
  • Rapid—entire purification protocol typically takes less than 40 mins
  • Economical—purification protocol allows multiple reuses
  • Versatile—beads are compatible with manual and automated workflows (e.g., KingFisher instruments)

Characteristics of Pierce Anti-DYKDDDDK Magnetic Agarose:

  • Composition: anti-DYKDDDDK antibody covalently attached to a magnetic, highly crosslinked agarose support
  • Magnetization: ferrimagnetic with low remanence
  • Bead size: 10–40 μm
  • Bead concentration: 25% slurry in phosphate buffered saline, 0.01% Tween-20 detergent, 0.02% sodium azide, pH 7.2
  • Binding capacity: ≥3.2 mg DYKDDDDK-tGFP-His protein (∼32 kDa)/mL settled beads
Specifications

Specifications

Magnetic Agarose
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Anti-DYKDDDDK Magnetic Agarose

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.