missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Antibody Clean-up Kit

Remove BSA, gelatin and other stabilizer proteins from antibody stock solutions before labeling and conjugation procedures.

Brand:  Thermo Scientific™ 44600

Product Code. 11885724

  • £320.00 / 10 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Antibody Clean-Up Kit is for removing BSA, gelatin and other stabilizer proteins from antibody stock solutions before labeling and conjugation procedures.

This is a complete and easy-to-use kit for removing BSA and gelatin stabilizing proteins (up to 1%) from IgG samples. The protocol uses Thermo Scientific Melon™ Gel to bind these frequently used storage proteins, allowing the antibody to flow through in a purified form. The antibody is recovered in an amine-free and carboxyl-free buffer, ready for use in downstream applications such as biotinylation, fluorescent labeling, crosslinking, immobilization and enzymatic fragmentation (e.g., Fab preparation). Bovine serum albumin (BSA) and gelatin are often added to purified antibody solutions as stabilizers for long-term storage. Inclusion of 0.2 to 1% (2 to 10mg/mL) BSA or gelatin helps stabilize dilute antibody solutions. Generally these added proteins do not interfere with immunodetection methods; however, their presence in solution does interfere with amine-reactive conjugation chemistries that are the basis for most biotinylation, fluorescent dye labeling, antibody fragmentation and covalent immobilization methods. For such methods, the antibody must be isolated from the stabilizing proteins, and the Antibody Clean-up Kit accomplishes this efficiently.

Highlights:

  • Fast – removes carrier protein from antibody solutions in half the time required for purifying antibodies through Protein A or G purification procedures

  • Gentle – no need to neutralize or desalt to mitigate the effects of harsh elution buffers

  • Complete – supplied desalting columns provide for easy buffer exchange into the required purification buffer or quick exchange into final buffer of choice

  • Economical – sufficient reagents to perform 10 antibody clean-up reactions

Includes:

Melon Gel IgG Purification Support and Buffer, desalting columns, microcentrifuge spin columns and tubes

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Antibody Clean-up Kit
Proprietary Resin, Affinity
Proprietary Ligand
Sufficient For: Purifying 10 x 0.5 mL IgG samples• Melon Gel IgG Purification Support, 1.2 mL
• Melon Gel Purification Buffer, 100 mL
• Zeba Spin Desalting Columns, 7K MWCO, 2 mL, 10 columns
• Spin Columns, 0.8 mL, 10 columns
• Microcentrifuge Tubes, 2 mL, 30 tubes

Store at 4°C
Pierce™
Kit, Microcentrifuge Spin Column
IgG Antibodies
10 Reactions
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZK402190 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number44600
Lot NumberZK402190A Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number44600
Lot NumberZH395953A Certificate TypeCertificate of Analysis Date04/09/2024 Catalog Number44600
Lot NumberZH395953 Certificate TypeCertificate of Analysis Date04/09/2024 Catalog Number44600
Lot NumberZF392039A Certificate TypeCertificate of Analysis Date11/07/2024 Catalog Number44600
Special Offers

Special Offers

  • Multiple Manufacturers
    Advance your proteomics research with essential tools
    Promo Code: 25977

    Expires2025-05-31

    Days

    58

    Hours

    23

    Minutes

    59

    Seconds

    44
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Antibody Clean-up Kit > Pierce™ Antibody Clean-up Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.