missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Jurkat Genomic DNA

Use this pure human genomic DNA as a control in DNA methylation analysis, genomic DNA library construction, PCR, and Southern blotting.

Brand:  Thermo Scientific™ SD1111

56.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 11921421

  • £78.75 / 15µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Jurkat Genomic DNA is human male genomic DNA from Jurkat cells. Genomic DNA was isolated from Jurkat E6.1 (human acute T-cell leukemia, clone E6.1) cells and grown to confluency in RPMI media supplemented with 10% bovine fetal serum. Isolated genomic DNA is dissolved in TE buffer.

Highlights:

  • Pure—DNA free from inhibitors, proteins, and RNA
  • Convenient—ready to use 100 ng/μL concentration
  • Compatible with downstream applications
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TE Buffer
Tube
[other]
15 μg
Liquid
100 ng/μL
Sequencing
Jurkat Genomic DNA
Human
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2806393 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberSD1111
Lot Number2794846 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberSD1111
Lot Number2832973 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberSD1111
Lot Number2852878 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberSD1111
Lot Number2907838 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberSD1111
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Jurkat Genomic DNA > 15μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.