Learn More
Thermo Scientific™ Pierce™ Boronic Acid Resin
Shop All Thermo Scientific ProductsDescription
Perform ribonucleotide and oligonucleotide RNA isolation with Thermo Scientific™ Pierce Boronic Acid Resin.
Highlights:
Ligand – boronic acid
Support – beaded polyacrylamide (1800 MW exclusion limit)
Spacer – m-aminophenyl group
Loading – 100μmol boronate per milliliter of settled resin
Capacity – at least 99% binding and recovery of 110μmol AMP per milliliter of settled resin
Format – 50% resin slurry in 0.05% sodium azide
Specifications
Specifications
| Content And Storage | Store at 4°C. |
| Form | Resin Slurry |
| Product Line | Pierce |
| Quantity | 10 mL |
| Target | Ribonucleotide |
| Type | Resin |
| Stationary Phase | Boronic Acid |
| Format | Bottle |
| Column Type | Affinity, Polyacrylamide Resin |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.