Learn More
Thermo Scientific™ CT(PEG)12 Carboxy-PEG-Thiol Compound
CT(PEG)n: carboxylic acid and sulfhydryl compound with a 12-unit polyethylene glycol (PEG) spacer; carboxylate and pegylate quantum dots, gold surfaces, silver.
Brand: Thermo Scientific™ 26133
Code : PN
Additional Details : Weight : 0.00100kg
Description
Thermo Scientific™ Pierce CT(PEG)12, or carboxy-PEG12-thiol, is a carboxyl- and sulfhydryl-terminated compound that contains a 12-unit polyethylene glycol (PEG) spacer and is used to modify surfaces such as quantum dots, monolayers and magnetic particles.
CT(PEG)12 is monodentate thiol-terminated and carboxylic acid pegylation reagent. The PEG compound has a defined molecular weight and spacer length and is useful for modifying surfaces such quantum dots, self-assembled monolayers and magnetic particles. Functionalization of solid surfaces with this polyethylene glycol reagent provides carboxylate “handles“ for protein immobilization via EDC crosslinking and significantly reduces nonspecific protein binding and provides.Highlights:
Metal-binding – terminal monodentate thiol reacts spontaneously with gold surfaces through dative binding
Carboxylate – terminal carboxylic acid group allows conjugation to amine-containing affinity ligands using carbodiimide (EDC) and Sulfo-NHS crosslinking chemistry
Polyethylene glycol – PEG groups are flexible, non-immunogenic, hydrophilic, and significantly reduce nonspecific binding of surfaces for protein methods
Spacer arm – 12-unit PEG spacer is nearly 48 angstroms long, minimizing steric hindrance for protein immobilization and binding assays
Specifications:
- Alternative name: Carboxy-PEG12-Thiol
- Chemical name: 1-mercapto-3,6,9,12,15,18,21,24,27,30,33,36-dodecaoxanonatriacontan-39-oic acid
- Chemical formula: C27H54O14S
- Molecular weight: 634.77
- Spacer arm: 47.8 angstroms
- Form: colorless to pale yellow liquid
- Solubility: dissolve in DMSO or DMF for use
Specifications
Amine | |
Conjugating Groups, Polyethylene Glycol (PEG) | |
12 ethylene glycol units | |
100 mg | |
Thiol Compound |
Store at -20°C protected from moisture. Shipped at ambient temperature. | |
Carboxylic Acid, Thiol | |
Solid | |
DMF or DMSO |
For Research Use Only. Not for use in diagnostic procedures.