missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ CDI-activated Agarose Resin

Covalently immobilize N-nucleophiles and primary amine ligands in aqueous or organic solvent conditions using carbonyldiimidazole-activated resin.

Brand:  Thermo Scientific™ 20259

Product Code. 10239963

  • £129.00 / 10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce CDI-Activated Agarose is a porous, beaded chromatography support that is activated with carbonyldiimidazole groups for covalent immobilization of amine-containing affinity ligands.

Pierce CDI-activated Agarose is a porous, beaded chromatography support that is activated with carbonyl diimidazole groups for covalent immobilization of proteins and other molecules that contain N-nucleophiles. Coupling can be performed in aqueous or organic solvent conditions, making possible the immobilization of many kinds of ligands. Once a molecule is immobilized, the prepared affinity resin can be used multiple times in typical affinity purification procedures.

Highlights:

  • Reliable coupling chemistry – immobilization occurs through the reaction of N-nucleophiles with 1,1'-carbonyldiimidazole groups of the resin to form stable, uncharged N-alkylcarbamate linkages

  • Easy to use – no secondary reagents needed; just wash equilibrate resin in alkaline coupling buffer and add ligand; reaction proceeds spontaneously

  • Stable activation – half-life of hydrolysis is much longer than hydroxysuccinimide ester activations, making immobilization reactions easier to prepare and control; simplifies filtration and washing before adding a ligand or protein

  • Well-defined coupling condition – reaction is most efficient with primary amines at pH 9 to 11

Properties of CDI-Activated Agarose:

  • Resin: crosslinked 6% beaded agarose
  • Activation: at least 50μmol 1,1'-carbonyldiimidazole (CDI) groups per milliliter of resin
  • Format: supplied as stabilized, 50% slurry in acetone
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at 4°C.
10 mL
Resin
Bottle
Pierce™
Primary Amine
Carbonyl Diimidazole
Affinity, Agarose Resin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.