Learn More
Thermo Scientific™ CHAPS Detergent (3-((3-cholamidopropyl) dimethylammonio)-1-propanesulfonate)
Non-denaturing Zwitterionic detergents that disrupt nonspecific protein interactions with less protein aggregation than nonionic detergents.
Brand: Thermo Scientific™ 28299
1482.00 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Thermo Scientific™ CHAPS is a zwitterionic detergent that is especially well suited for protecting the native state of proteins.
CHAPS is a sulfobetaine derivatives of cholic acid. This zwitterionic detergent is useful for membrane protein solubilization when it is important to maintain protein activity. CHAPS detergent is soluble over a wide range of pH (2 to 12) and are easily removed from solution by dialysis because of their high critical micelle concentration (CMC).
Highlights:
- Versatile zwitterionic detergents
- Provide mild but effective lysis of cultured mammalian cells
- Nondenaturing and generally do not inactivate protein functions
- Can be removed from solution by dialysis
- Commonly used for isoelectric focusing (IEF) and 2D electrophoresis
Properties of CHAPS detergent:
- Chemical Name: 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate
- Molecular Weight: 614.88
- Detergent Class: Zwitterionic
- Aggregation Number: 10
- Micelle Molecular Weight: 6149
- Critical Micelle Concentration (CMC): 8 to 10mM (0.4920 to 0.6150%, w/v)
- Cloud Point: ≥ 100°C
- Dialyzable: Yes

Specifications
Detergent | |
Powder | |
100 g |
Store at Room Temperature | |
Detergent (Pure) |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Powering Proteomics
Advance your proteomics research with essential toolsPromo Code: 25977Expires: 2025-05-31
New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.