missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Chicken IgY Purification Kit

Purify chicken antibodies from hen egg yolks with a unique delipidation reagent and precipitation reagent to selectively isolate the IgY fraction.

Brand:  Thermo Scientific™ 44918

 View more versions of this product

Product Code. 10006433

  • £304.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Chicken IgY Purification Kit purifies chicken antibodies from hen egg yolks using a unique delipidation reagent to separate protein from lipid and a precipitation reagent to selectively isolate the IgY fraction.

The IgY Purification Kit is simple and easy to use. After isolating a yolk using the supplied egg separator, lipids are effectively removed by incubation and centrifugation in Pierce Delipidation Reagent. This first reagent is also effective as a storage solution, enabling multiple yolks to be collected and stored for simultaneous extraction at a later date. Upon addition of the specialized Pierce IgY Precipitation Reagent and subsequent centrifugation, a nearly pure pellet of intact IgY results; it is easily dissolved in phosphate buffered saline (PBS) or other storage buffer. The purified IgY is ready for use in immunodetection methods or antigen-specific purification without interference from lipids.

Highlights:

  • Cost-effective – purifies twice as much IgY per egg than other popular kits, providing a lower cost-per-milligram of purified IgY

  • High purity – method yields chicken antibody that is 85 to 95% pure (based on SDS-PAGE analysis)

  • Easy to use – simple precipitation method purifies chicken IgY without affinity columns

  • Flexible protocol – eggs can be harvested and quickly stored in buffer for purification at a later date

  • Convenient – use eggs directly out of refrigerator; no need to equilibrate them to room temperature

Includes:

Egg separator, delipidation reagent and IgY precipitation reagent

TRUSTED_SUSTAINABILITY
Specifications

Specifications

IgY Purification Kit
Precipitation
IgY Antibody
1 L
Pierce™
Kit, Bottle
Sufficient For: Purifying chicken IgY from 5 egg yolks
• Delipidation Reagent, 500 mL
• IgY Precipitation Reagent, 500 mL
• Egg Separator, 1 separator

Store at 4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.