Learn More
Thermo Scientific™ Pierce™ Chicken IgY Purification Kit
Purify chicken antibodies from hen egg yolks with a unique delipidation reagent and precipitation reagent to selectively isolate the IgY fraction.
£304.00 - £1068.00
Specifications
Product Type | IgY Purification Kit |
---|---|
Product Line | Pierce™ |
Column Type | Precipitation |
Format | Kit, Bottle |
Purification Target | IgY Antibody |
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
10006433
|
Thermo Scientific™
44918 |
Pierce Chicken IgY Purification Kit | 1 L |
£304.00
1 set |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10559673
|
Thermo Scientific™
44922 |
Pierce Chicken IgY Purification Kit | 5 L |
£1068.00
1 set |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Thermo Scientific™ Pierce Chicken IgY Purification Kit purifies chicken antibodies from hen egg yolks using a unique delipidation reagent to separate protein from lipid and a precipitation reagent to selectively isolate the IgY fraction.
The IgY Purification Kit is simple and easy to use. After isolating a yolk using the supplied egg separator, lipids are effectively removed by incubation and centrifugation in Pierce Delipidation Reagent. This first reagent is also effective as a storage solution, enabling multiple yolks to be collected and stored for simultaneous extraction at a later date. Upon addition of the specialized Pierce IgY Precipitation Reagent and subsequent centrifugation, a nearly pure pellet of intact IgY results; it is easily dissolved in phosphate buffered saline (PBS) or other storage buffer. The purified IgY is ready for use in immunodetection methods or antigen-specific purification without interference from lipids.Highlights:
Cost-effective – purifies twice as much IgY per egg than other popular kits, providing a lower cost-per-milligram of purified IgY
High purity – method yields chicken antibody that is 85 to 95% pure (based on SDS-PAGE analysis)
Easy to use – simple precipitation method purifies chicken IgY without affinity columns
Flexible protocol – eggs can be harvested and quickly stored in buffer for purification at a later date
Convenient – use eggs directly out of refrigerator; no need to equilibrate them to room temperature
Includes:
Egg separator, delipidation reagent and IgY precipitation reagent
Specifications
IgY Purification Kit | |
Precipitation | |
IgY Antibody |
Pierce™ | |
Kit, Bottle | |
Pierce Chicken IgY Purification Kit |
For Research Use Only. Not for use in diagnostic procedures.