Learn More
Thermo Scientific™ Pierce™ Control Agarose Resin
Unmodified beaded agarose resin for pre-clearing protein immunoprecipitation samples to reduce background signal and nonspecific binding in IP assays.
Brand: Thermo Scientific™ 26150
Code : PN
Additional Details : Weight : 0.68040kg Transport : UN number : 3276 Chem class : 6.1 Pack group : III
Description
Thermo Scientific™ resin is effective for depleting proteins and other sample components that bind to agarose nonspecifically, which reduces background signal in IP reactions
When used as a negative control, these beads enable specific and nonspecific binding interactions to be distinguished.Highlights:
- Crosslinked 4% beaded agarose (CL4B) is the same base support used in various Thermo Scientific Pierce IP and co-IP kits, unmodified and non-activated
- 10mL package size is more appropriate for experimental needs
- Supplied as a 50% slurry in 20% ethanol (pipetting 1mL of slurry delivers 0.5mL of settled resin): wash in buffer and the beads are ready to use
Specifications
Store at 2–8°C. | |
Pierce™ | |
Resin | |
Agarose Resin |
Liquid Suspension | |
10 mL | |
Bottle |
Safety and Handling
- Control Agarose Resin
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
For Research Use Only. Not for use in diagnostic procedures.