Learn More
Thermo Scientific™ Pierce™ Cypridina Luciferase Flash Assay Kit
Assay regulatory element transcriptional activity in mammalian cell culture media and whole cell lysate with this highly sensitive flash-based luciferase kit.
Brand: Thermo Scientific™ 16169
398.68 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
The Thermo Scientific™ Pierce Cypridina Luciferase Flash Assay Kit provides researchers with a highly sensitive assay for transcriptional activity of regulatory elements in mammalian cell culture media and whole cell lysate.
This Flash Assay Kit contains reagents for measuring the activity of Cypridina luciferase in mammalian cell culture media and lysates. When used with Thermo Scientific Cypridina Luc Vectors, the kit provides an extremely sensitive bioluminescent reporter assay system for secreted or intracellular detection of promoter or pathway activity. Cypridina luciferase is highly secreted into the cell culture media, allowing for live cell monitoring of reporter activity. The signal produced by Cypridina is considerably greater than signal from either firefly or Renilla luciferases assayed under similar conditions.
Highlights:
- Sensitive – high sensitivity allows utilization of smaller numbers of cells
- Cost effective – sensitive assays result in decreased reagent consumption
- Secreted luciferase – allows real-time assays and kinetic studies without destroying cells
- Ideal for co-transfection – uses a different substrate than Renilla, Gaussia, or firefly luciferases
- Compatible – assay reagents compatible with other Cypridina or Vargula luciferases
- Time-saving – assays using secreted luciferases require minimal sample handling
- Automation-friendly – amenable to high throughput screens
- Convenient – contains a universal cell lysis buffer and optimized flash assay reagent
- Safe – allows one to perform non-radioactive assays
Includes:
Cell lysis buffer, reaction buffer and substrate
Requires:
Cypridina luciferase and luminometer or other instrument capable of monitoring luminescence, such as Thermo Scientific Luminoskan Ascent and Varioskan Flash Microplate Readers.
Recommended for:
Promoter studies for analyzing cis- regulatory elements and trans-acting factors; Drug screening; siRNA and miRNA screening; Multiplexed assays to study off-target effects; Secretory pathway / protein localization reporter assays; Signal transduction pathway analysis; RNA splicing studies
Specifications
Assay Kit | |
1000 Reaction Kit | |
Reporter Enzyme, Luciferase Reporter Assay | |
Bioluminescence | |
384-well plate, 96-well plate | |
Chemical Substrate | |
Luciferase Substrate | |
Vargulin (Cypridina Luciferin) |
Luciferase, Cypridina Luciferase | |
• Cypridina Flash Assay Buffer, 5 mL, store at 4°C • Vargulin (1X), 0.5 mL, store at -20°C • 2X Cell Lysis Buffer, 6 mL, store at room temperature Upon receipt store kit at -20°C or store individual components as indicated above. |
|
Mammalian Cells | |
Luminometer (Microplate) | |
Enzyme Labeled | |
Enhanced Chemiluminescence | |
Pierce™ |
For Research Use Only. Not for use in diagnostic procedures.