Learn More
Thermo Scientific™ Pierce™ Disposable Columns, 5 mL
Empty drip columns (0.5 to 5 mL) with matching frits and caps for packing beaded affinity or desalting resins to use in gravity-flow purification techniques.
Brand: Thermo Scientific™ 29922
297.35 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : PN
Additional Details : Weight : 1.81440kg
Description
Thermo Scientific™ Pierce Disposable Plastic Columns are empty columns with matching frits and caps for use in a variety of resin-based affinity or desalting techniques.
Additional Information and Online Ordering »
Highlights:
- For bench top gravity-flow affinity purification and size exclusion chromatography
- Supplied complete with porous polyethylene discs, stoppers and end caps
- Columns can be pre-packed and stored until needed
- Compatible with most types of aqueous buffer eluents commonly used in chromatography
- Plastic funnels can be attached to expand the buffer reservoir for increased flow rates
Includes:
Trial pack contains two columns of each available size (six columns total), matching column accessories (discs and caps), and one funnel (column reservoir extender)
Specifications
5 mL | |
Store at room temperature. | |
Spin Column | |
Empty Column, Polyethylene Filter |
Disposable Columns, 5mL | |
Pierce™ | |
Gravity Flow Column |
Safety and Handling
- Pierce Disposable Columns, 5 mL
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : thylbutyl)phenyl ]--hydroxy poly(oxy-1,2-ethanediyl)
For Research Use Only. Not for use in diagnostic procedures.