missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ Disposable Columns, 10 mL
Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 columns
Unit Size:
Pack of 100
Description
Thermo Scientific™ Pierce Disposable Plastic Columns are empty columns with matching frits and caps for use in a variety of resin-based affinity or desalting techniques.
Additional Information and Online Ordering
These bulk packages of empty columns are economical and convenient devices for packing affinity purification supports and desalting resins for gravity-flow procedures. Three columns sizes are offered, suitable for procedures requiring 0.5 to 10mL resin-bed volumes. The plastic columns are supplied with matching pairs of polyethylene discs and polypropylene caps (top and bottom). The columns are economically-priced for single use and sufficiently durable to prepare and store columns for multiple uses.
Highlights:
- For bench top gravity-flow affinity purification and size exclusion chromatography
- Supplied complete with porous polyethylene discs, stoppers and end caps
- Columns can be pre-packed and stored until needed
- Compatible with most types of aqueous buffer eluents commonly used in chromatography
- Plastic funnels can be attached to expand the buffer reservoir for increased flow rates
Includes:
Trial pack contains two columns of each available size (six columns total), matching column accessories (discs and caps), and one funnel (column reservoir extender)
Specifications
Specifications
| Capacity (Metric) | 10 mL |
| Description | Disposable Columns, 10mL |
| Content And Storage | Store at room temperature. |
| Product Line | Pierce |
| Quantity | 100 columns |
| Type | Spin Column |
| Format | Gravity Flow Column |
| Column Type | Empty Column, Polyethylene Filter |
Safety and Handling
Product Identifier
- Disposable Polypropylene Columns
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : hylbutyl)phenyl]-ydroxy-Poly(oxy-1,2-ethanediyl)
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction