missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ DMP (dimethyl pimelimidate)
Crosslink amines to amines with dimethyl pimelimidate (CAS 58537-94-3); composed of imidoester groups and a membrane-permeable, 7-atom spacer arm.
Brand: Thermo Scientific™ 21667
70.06 GBP valid until 2024-12-31
Use promo code "21745" to get your promotional price.
This item is not returnable.
View return policy
Description
Thermo Scientific™ Pierce DMP is dimethyl pimelimidate, a membrane-permeable crosslinker that contains an amine-reactive imidoester group at each end of a 7-atom spacer arm.
Highlights:
- Water-soluble
- Rapid reaction with amines at alkaline pH values (pH 8-10)
- Amidine bond retains net charge character of protein
- Reversible at high pH values
- Reactive groups: imidoester (homobifunctional)
- Reactive toward: amino groups
Specifications
Amine-Amine | |
DMP | |
No | |
Upon receipt store desiccated at 4°C. | |
Ambient | |
Chemical Labeling | |
Imidoester | |
Powder | |
Water | |
Yes |
No | |
259.17 | |
9.2 Å | |
Yes | |
Pierce™ | |
Homobifunctional | |
Short (<10 Å) | |
1 g | |
Standard | |
Crosslinker |
For Research Use Only. Not for use in diagnostic procedures.