Learn More
Thermo Scientific™ Pierce™ DSP, No-Weigh™ Format
Crosslink amines to amines with dithiobis succinimidyl propionate, composed of NHS-ester groups and disulfide-containing, 12.0A spacer arm and cleavable with reducing agents.
Brand: Thermo Scientific™ 22586
148.64 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : PN
Additional Details : CAS Number : 57757-57-0 Weight : 0.45360kg
Description
Thermo Scientific™ Pierce DSP Crosslinker (Lomant's Reagent) is an amine-reactive crosslinker with NHS-ester reactive ends and a cleavable disulfide bond in its spacer arm.
Highlights:
- Cleavable by reducing agents
- Membrane-permeable
- Acylate in aqueous or organic media within two minutes at room temperature
- Four- to five-hour half-life of ester at pH 7.0
- Reactive groups: NHS ester (homobifunctional)
- Reactive toward: amino groups
Specifications
Amine-Amine | |
DSP | |
No | |
Upon receipt store desiccated at 4°C. | |
Ambient | |
Chemical Labeling | |
NHS Ester | |
Powder | |
DMF, DMSO | |
No |
By Thiols | |
404.42 | |
12.0 Å | |
Yes | |
Pierce™ | |
Homobifunctional | |
Medium (10 to 30 Å) | |
50 mg | |
Standard, Single-use, Premium-grade | |
Crosslinker |
Safety and Handling
- DSP (dithiobis(succinimidyl propionate)), Lomant's Reagent
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : disuccinimido dithiobispropionate
For Research Use Only. Not for use in diagnostic procedures.