missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ DTT (Dithiothreitol)
Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 g
Unit Size:
5g
Description
Dithiothreitol is a popular reducing agent for many protein research applications, including reducing polyacrylamide electrophoresis (SDS-PAGE).
Dithiothreitol is a popular reducing agent for many protein research applications, including reducing polyacrylamide electrophoresis (SDS-PAGE). 5g glass bottle.
Specifications
Specifications
| Description | Pierce DTT, Cleland's Reagent |
| Content And Storage | Store at 4°C. |
| Product Line | Pierce |
| Form | Powder |
| Quantity | 5 g |
| Product Type | Dithiothreitol |
Safety and Handling
Product Identifier
- DITHIOTHREITOL (DTT,CLELAND'S REAGENT)5 GM
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- MIXTURE LIST-Contains : 1,4-dimercapto-2,3-Butanediol
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction