Learn More
Thermo Scientific™ Pierce™ ECL Plus Western Blotting Substrate
Acridan-based chemiluminescent and chemifluorescent HRP substrate for Western blot detection using X-ray film or CCD- or laser-based imagers.
Brand: Thermo Scientific™ 32132
183.61 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
Thermo Scientific™ Pierce ECL Plus Substrate is an acridan-based chemiluminescent and chemifluorescent substrate to detect peroxidase activity from HRP-conjugated probes by Western blotting using X-ray film, CCD or laser-based imagers.
Pierce ECL Plus Substrate is a chemiluminescent and chemifluorescent substrate to detect peroxidase activity from HRP-conjugated antibodies and probes by Western blotting using X-ray film, CCD camera documentation systems or laser-based imagers. Pierce ECL Plus Substrate generates acridinium esters when it reacts with HRP. As these ester intermediates react with peroxide, they produce a strong and sustained chemiluminescent signal that can be captured by X-ray film CCD imaging. The substrate also produces a robust fluorescent signal that can be captured with fluorescence imagers. Pierce ECL Plus Substrate enables the detection of low-picogram amounts of target protein on nitrocellulose or PVDF membrane when probed with appropriate primary and secondary antibody concentrations. Its broad dynamic range, high sensitivity and long-lasting signal make Pierce ECL Plus Substrate an excellent choice for Western blotting.Highlights:
Easy to use — can be substituted for the discontinued GE ECL Plus Substrate without any re-optimization
Higher sensitivity — detect targets down to the low-picogram level
Longer signal duration — sustained light output for as long as five hours
More imaging options — X-ray, CCD or laser-based imagers
More affordable — high quality and performance at a lower price than other suppliers« similar substrates
Specifications
Pierce ECL Plus Western Blotting Substrate | |
Chemiluminescence, Fluorescence | |
Protein Assay and Analysis | |
Pierce™ | |
HRP | |
10 mini-gel blots; 1000cm2 of membrane | |
2°C to 8°C |
Substrate | |
Liquid | |
Nitrocellulose, PVDF | |
100 mL Kit | |
Low picogram to mid-femtogram | |
Detection Reagent A, 100mL, Detection Reagent B, 2.5mL |
Safety and Handling
- Pierce ECL Plus Western Blotting Substrate
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
Precautionary Statement
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
- MIXTURE LIST-Contains : ethanol, 1,4-dioxane
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.