missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ ECL Plus Western Blotting Substrate

Acridan-based chemiluminescent and chemifluorescent HRP substrate for Western blot detection using X-ray film or CCD- or laser-based imagers.

Brand:  Thermo Scientific™ 32132X3

653.65 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11527271

  • £718.00 / Pack of 3

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce ECL Plus Substrate is an acridan-based chemiluminescent and chemifluorescent substrate to detect peroxidase activity from HRP-conjugated probes by Western blotting using X-ray film, CCD or laser-based imagers.

Thermo Scientific Pierce ECL Plus Substrate is an acridan-based chemiluminescent and chemifluorescent HRP substrate for western blot detection using X-ray film or CCD- or laser-based imagers. This product is sold as Pierce ECL 2 Substrate (Cat. #PI80196X3) through Fisher Scientific and other channels.

Features of ECL Plus Substrate:

  • Easy to use—can be substituted for the discontinued GE ECL Plus Substrate without any re-optimization
  • Higher sensitivity—detect targets down to the low-picogram level
  • Longer signal duration—sustained light output for as long as 5 hours
  • More imaging options—X-ray, CCD or laser-based imagers
  • More affordable—high quality and performance at a lower price than other suppliers' similar substrates

    Pierce ECL Plus Substrate generates acridinium esters when it reacts with HRP. As these ester intermediates react with peroxide, they produce a strong and sustained chemiluminescent signal that can be captured by X-ray film CCD imaging. The substrate also produces a robust fluorescent signal that can be captured with fluorescence imagers. Pierce ECL Plus Substrate enables the detection of low-picogram amounts of target protein on nitrocellulose or PVDF membrane when probed with appropriate primary and secondary antibody concentrations. Its broad dynamic range, high sensitivity and long-lasting signal make Pierce ECL Plus Substrate an excellent choice for western blot analysis.
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Substrate
    Liquid
    Nitrocellulose, PVDF
    300 mL Kit
    3000 sq. cm of membrane
    Chemiluminescence, Fluorescence
    Western Blot
    Pierce™
    Low picogram to mid-femtogram
    Upon receipt store at 4°C. Product shipped on ice.
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

    For Research Use Only. Not for use in diagnostic procedures.