missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Fab Preparation Kit

Digest human or mouse IgG antibodies with immobilized papain protease and purify Fab fragments using Protein A agarose.

Brand:  Thermo Scientific™ 44985


Product Code. 10269923

  • £710.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Pierce Fab Preparation Kit
10 Antibody Samples, each containing 0.25 to 4 mg IgG kit
Pierce™
Sufficient For: 10 antibody samples, each containing 0.25 to 4 mg IgG
• Pierce Papain Agarose, 1.25 mL
• Cysteine-HCl, 1 g
• Fab Digestion Buffer, 120 mL
• NAb Protein A Plus Spin Column, 1 mL, 1 column
• PBS Packs (each makes 500 mL), 2 packs
• IgG Elution Buffer, 120 mL
• Zeba Desalt Spin Columns, 7K MWCO, 2mL, 10 columns
• Spin Columns, 0.8 mL, 10 columns
• Microcentrifuge Tubes, 2 mL, 30 tubes

Store at 4–8°C.
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberSH252820 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number44985
Lot NumberZK401608 Certificate TypeCertificate of Analysis Date18/12/2024 Catalog Number44985
Lot NumberZK401608A Certificate TypeCertificate of Analysis Date18/12/2024 Catalog Number44985
Lot NumberZJ398034 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number44985
Lot NumberZK399250 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number44985
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Fab Preparation Kit > Kit for 0.25 to 4mg IgG samples; 1.25mL-resin kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.