Learn More
Thermo Scientific™ Pierce™ Glutathione Superflow Agarose
Perform high-purity, large-scale purification of GST-tagged proteins with glutathione that has been immobilized to a crosslinked 6% beaded agarose.
£272.00 - £8400.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13434199
|
Thermo Scientific™
25237 |
50 mL |
£840.00
50mL |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
13444199
|
Thermo Scientific™
25239 |
1 L |
£8400.00
1 L |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
13424199
|
Thermo Scientific™
25236 |
10 mL |
£272.00
10mL |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
13454199
|
Thermo Scientific™
25238 |
250 mL |
£2790.00
250mL |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
Description
Perform high-purity, large-scale purification of GST-tagged proteins with Thermo Scientific™ Pierce Glutathione Superflow Agarose, which consists of glutathione that has been immobilized to a crosslinked 6% beaded agarose.
Additional Information and Online Ordering »
Highlights:
High dynamic binding capacity – binds 10mg of pure GST per mL of resin at a linear flow rate of 150cm/hour; can bind greater than 20mg/mL at 30cm/hour flow rate
High purity – provides greater than 90% purity when used to purify overexpressed GST or GST-fusion proteins from lysates
Robust – highly crosslinked 6% agarose Superflow beaded support tolerates linear flow rates up to 1200cm/hour
Compatible – maintains function after exposure to a wide variety of chemicals and pH values
Cost effective – resin is stable through multiple cycles of cleaning and reuse
Recommended for:
Medium- to large-scale FPLC purification of GST-tagged proteins
For Research Use Only. Not for use in diagnostic procedures.