missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ High Capacity Endotoxin Removal Resin

Remove endotoxins (lipopolysaccharides or LPS) from samples before mass spectrometry using spin columns of poly(e-lysine) ligand on beaded cellulose.

Brand:  Thermo Scientific™ 88272

3649.40 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 12331973

  • £5140.00 / 250mL
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Reduce endotoxin levels in protein samples by greater than 99% in one hour while maximizing protein recovery with Thermo Scientific™ Pierce High Capacity Endotoxin Removal Resin.

The Pierce High Capacity Endotoxin Removal Resin combines porous cellulose beads and an FDA-approved food preservative, poly(epsilon-lysine), as an affinity ligand to selectively bind endotoxins. Endotoxin levels in protein samples are reduced by greater than 99% in as little as 1 hour using our spin cup format, and protein recovery is ≥85%. The modified polylysine affinity ligand eliminates the toxicity concerns associated with alternative technologies using polymixin B ligands and sodium deoxycholate buffers. Pierce High capacity Endotoxin Removal Resin is available as a slurry to pack a custom column that can be re-used at least 5 times with no loss in endotoxin removal efficiency. This resin is also available in a new single-use spin column format optimized for different sample volumes.

Highlights:

  • High capacity bind up to 2,000,000 EU/mL to eliminate at least 99% of endotoxins

  • Durable re-use resin up to 10 times with no loss in performance

  • Selective recover at least 85% of your protein sample

  • High performance compatible with FDA guidelines by reducing final EU concentration to less than 5 EU/mL

  • Fast our new spin cup format enables endotoxin depletion within 1 hour

  • Cleansingle-use spin columns avoid cross contamination of samples

  • Optimized spin cups are optimized for different sample volumes

  • Economical large-volume discounts available

Requires:

Endotoxin-free water

Recommended for:

Eliminate endotoxins from protein or antibody samples prior to cell transfection or animal injection

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Up to 2,000,000 EU/mL
Upon receipt store at 4°C.
250 mL
Resin
Bottle
Proprietary ligand on beaded cellulose slurried in 20% ethanol
Pierce™
Endotoxin Removal
Proprietary Ligand
Affinity, Proprietary Resin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.