Learn More
Thermo Scientific™ Pierce™ High Capacity Endotoxin Removal Spin Columns, 0.5 mL
Remove endotoxins (lipopolysaccharides or LPS) from samples before mass spectrometry using spin columns of poly(e-lysine) ligand on beaded cellulose.
Brand: Thermo Scientific™ 88275
209.45 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
Reduce endotoxin levels in protein samples by greater than 99% in one hour while maximizing protein recovery with Thermo Scientific™ Pierce High Capacity Endotoxin Removal Resin.
The Pierce High Capacity Endotoxin Removal Resin combines porous cellulose beads and an FDA-approved food preservative, poly(epsilon-lysine), as an affinity ligand to selectively bind endotoxins. Endotoxin levels in protein samples are reduced by greater than 99% in as little as 1 hour using our spin cup format, and protein recovery is ≥85%. The modified polylysine affinity ligand eliminates the toxicity concerns associated with alternative technologies using polymixin B ligands and sodium deoxycholate buffers. Pierce High capacity Endotoxin Removal Resin is available as a slurry to pack a custom column that can be re-used at least 5 times with no loss in endotoxin removal efficiency. This resin is also available in a new single-use spin column format optimized for different sample volumes.Highlights:
High capacity bind up to 2,000,000 EU/mL to eliminate at least 99% of endotoxins
Durable re-use resin up to 10 times with no loss in performance
Selective recover at least 85% of your protein sample
High performance compatible with FDA guidelines by reducing final EU concentration to less than 5 EU/mL
Fast our new spin cup format enables endotoxin depletion within 1 hour
Cleansingle-use spin columns avoid cross contamination of samples
Optimized spin cups are optimized for different sample volumes
Economical large-volume discounts available
Requires:
Endotoxin-free water
Recommended for:
Eliminate endotoxins from protein or antibody samples prior to cell transfection or animal injection
Specifications
Up to 2,000,000 EU/mL | |
Upon receipt store at 4°C. | |
Pierce™ | |
Endotoxin Removal | |
Proprietary Ligand | |
Affinity, Cellulose Resin |
8mL-capacity centrifuge columns containing 0.5mL resin | |
Liquid | |
25 Columns | |
Spin Column | |
Spin Column |
Safety and Handling
- Pierce High Capacity Endotoxin Removal Spin Columns, 0.5 mL
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
Hazard Statement
- H228-Flammable solid.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.