missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ High Capacity Ni-IMAC Resin, EDTA compatible

Pierce high-capacity EDTA-compatible Ni-IMAC resin is designed for the purification of over-expressed His-tagged proteins from cell culture media.

Brand:  Thermo Scientific™ A50586

 View more versions of this product

Product Code. 17193259

  • £2165.00 / 250mL
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Pierce high capacity Ni-IMAC resin contains a matrix that is a 6% crosslinked agarose with optimal properties for batch, gravity column, and research/pilot scale FPLC column purifications. The resin has high binding capacity (up to 80 mg of protein per mL of resin) and can tolerate the high EDTA and reducing agent concentrations used to inhibit metalloproteases and stabilize sensitive intracellular proteins.

Features of the high-capacity EDTA-compatible Ni-IMAC resin include:

  • Chelator stable—use up to 20 mM EDTA and DTT with no loss in performance
  • Compatible with purification of overexpressed, secreted proteins in cell culture media (e.g., Expi293, ExpiCHO, and ExpiSf expression systems)
  • High binding capacity—>80 mg/mL binding capacity for His-tagged recombinant proteins
  • Reusable—can be regenerated up to 10 times using the CIP protocol

EDTA and reducing agents such as DTT and TCEP adversely affect the performance of Ni-IMAC supports by stripping off the metal. Because of its chelator stability characteristic (EDTA tolerant), Pierce high-capacity EDTA-compatible Ni-IMAC resin is especially suited for purifying expressed His-tagged proteins that are secreted into cell culture media or for purifying intracellular His-tagged proteins that need the presence of EDTA to maintain stability and function. In both cases, large quantities of samples can be purified without the Ni metal stripping off of the support, thereby providing consistent high capacity yields.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Chelator Stable
250 mL settled resin, supplied as 500 mL 50% slurry. Store at 2–8°C.
Pierce™
Resin
6% crosslinked agarose
Wet ice
250 mL
Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.