Learn More
Thermo Scientific™ Pierce™ High Capacity Protein A MagBeads, alkali stable
The Pierce High-Capacity Protein A MagBeads, Alkali Stable are a high-performance magnetic bead for purifying antibodies from cell culture supernatant, ascites, serum, and crude extracts.
Brand: Thermo Scientific™ A53038
Additional Details : Weight : 0.50000kg
Description
- High-performance magnetic beads for purifying antibodies from cell culture supernatant, ascites, serum, and crude extracts
- The ligand is produced in yeast and is immobilized on magnetic agarose with an average particle size of ∼30 mm
- Engineered to withstand standard CIP procedures with 0.1M NaOH, enabling efficient cleaning of the beads for reuse
- Beads can be regenerated at least 10 times without sacrificing binding capacity
- Suitable for purifying mAbs and a subpopulation of scFv’s and Fab fragments containing a VH3 domain
- Compatible with manual and automated workflows (e.g., KingFisher instruments)
- High-affinity Protein A is securely immobilized to magnetic agarose to prevent leaching of ligand, target antibody, or base support
- High binding capacity of ≥ 40 mg HIgG/mL settled beads
- Animal component-free raw materials
Specifications
≥40 mg HIgG/mL settled beads | |
Provided in the form of a 25% slurry. Upon receipt store at 2–8°C. |
|
4 x 25 mL | |
Alkaline Stable Protein A |
High Capacity Magnetic Agarose Beads | |
∼ 30 μm | |
Antibody Purification | |
Bottle |
For Research Use Only. Not for use in diagnostic procedures.