Learn More
Thermo Scientific™ Pierce™ HRV 3C Protease Solution Kit (2 units/μL)
Remove fusion tags from proteins with the HRV 3C cleavage sequence with this recombinant cysteine protease that is dual tagged for easy removal from the sample after cleavage.
£309.00 - £2520.00
Specifications
Content And Storage | Store at -20°C. |
---|---|
Product Line | Pierce™ |
Form | Kit |
Product Type | Protease |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13483689
|
Thermo Scientific™
88946 |
1000 U |
£309.00
1000 units |
In Stock
Log in to see stock. |
|||||
13454289
|
Thermo Scientific™
88947 |
10,000 U |
£2520.00
10ku |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
Description
Remove fusion tags from proteins with the HRV 3C cleavage sequence with Thermo Scientific™ Pierce Human Rhinovirus (HRV) 3C Protease, a recombinant cysteine protease that is dual tagged for easy removal from the sample after cleavage.
Additional Information and Online Ordering »
Highlights:
Effective – HRV 3C is designed to remove purification tags from your protein of interest
High activity at 4°
C – activity is ≥2000U/mg control protein; one unit will cleave 0.1mg control protein at 4°C after 16h
Flexible – dual-tagged enzyme is compatible with various columns and buffers
Includes:
Kit contains protease, cleavage buffer and control protein.
Recommended for:
Removal of purification tags from fusion proteins
Specifications
Store at -20°C. | |
Pierce™ | |
Kit | |
Protease |
For Research Use Only. Not for use in diagnostic procedures.