missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protein G IgG Plus Orientation Kit, 2 mL

Product Code. 10289923 Shop All Thermo Scientific Products
Click to view available options
Quantity:
2 Columns
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 10289923

Brand: Thermo Scientific™ 44990

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bind and covalently immobilize antibodies to create a reusable affinity purification column with high binding capacity with Protein A or G agarose columns and DSS crosslinker.

Thermo Scientific™ Pierce IgG Plus Orientation Kits bind and crosslink antibody to Protein A or Protein G agarose resin, thereby creating a reusable affinity column for purification of antigens.

These IgG orientation kits use a crosslinker to covalently attach antibody that is bound to Protein A or Protein G agarose resin, thereby creating a reusable affinity column for purification of antigens. The Plus Orientation Kits provide for more efficient and stable antibody immobilization than the traditional orientation method because they use the NHS-ester crosslinker DSS (disuccinimidyl suberate) rather than the imidoester crosslinker DMP (dimethyl pimelimidate). Several milligrams of IgG can be bound and crosslinked to each 2mL column, afterwhich the prepared affinity column can be used multiple times for antigen purification using typical binding and elution conditions.

The immobilization method is called orientation because antibodies are first captured to the affinity resin through their Fc regions, which are the target binding sites of Protein A and Protein G ligands. Thus, the antibodies are bound with their antigen-binding sites oriented outward. DSS crosslinker reacts with primary amines (side chain of lysine residues) of the Protein A/G and affinity-bound antibody, thereby covalently and permanently attaching the proteins together.

Highlights:

  • Contaminant-free antigen purification – enables antigen purification without contamination (co-elution) of antibody

  • Covalent, leak-resistant antibody immobilization – high-capacity binding of IgG to Protein A or Protein G agarose, followed by efficient crosslinking with DSS crosslinker

  • High-quality agarose resin – crosslinked 6% beaded agarose with recombinant Protein A or Protein G

  • Gentle binding and crosslinking conditions – coupling antibody in standard PBS

  • Oriented immobilization – binds and crosslinks IgG with the antigen-binding sites directed outward, able to capture passing antigen in the mobile phase without steric hindrance

  • Complete kits – include all the reagents needed to immobilize the antibody

  • Better than DMP – less antibody leaching than traditional orientation method with dimethyl pimelimidate crosslinker

Includes:

Protein A or G columns, disuccinimidyl suberate (DSS), PBS pack, blocking buffer, binding/wash buffer, elution buffer, and column accessories

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 2 mL
Content And Storage Sufficient For: Immobilizing 1 to 6 mg of mouse IgG per column
• Protein G Columns, 2 mL, 2 columns
• Disuccinimidyl Suberate (DSS), 2 x 13 mg
• PBS Pack (makes 500 mL), 1 pack
• Blocking Buffer, 6 mL
• IgG Elution Buffer, 15 mL
• Antibody Binding/Wash Buffer, 120 mL
• Column Accessories

Store at 4°C.
Product Line Pierce
Quantity 2 Columns
Type IgG Plus Orientation Kit
Stationary Phase Protein G
Sufficient For 1 to 6mg Mouse IgG per Column
Format Spin Column
Column Type Affinity, Agarose Resin

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.