Learn More
Thermo Scientific™ Pierce™ Mannan Binding Protein Agarose
Easily purify IgM from mouse ascites fluid using immobilized mannan binding protein, which binds to IgM in the kit-supplied buffers.
Brand: Thermo Scientific™ 22212
Description
Thermo Scientific™ Pierce IgM Purification Kit is an easy-to-use affinity matrix and buffer system to purify IgM from mouse ascites fluid; the method uses immobilized mannan binding protein, which binds to IgM in the kit-supplied buffers.
Pierce MBP Agarose is purified mannan binding protein (also called mannose-binding lectin, MBL) that has been immobilized on 4% beaded agarose. When used with an optimized buffer system based on temperature- and calcium-dependent binding, this resin enables specific affinity purification of mouse IgM from ascites fluid and other concentrated samples. The resin and essential buffers are available individually or bundled together in a complete IgM Purification Kit containing a pre-packed 5mL column of the MBP Agarose.
Highlights:
Immobilized MBP – affinity system based on mannan binding protein (MBP) allows specific purification of mouse IgM from ascites fluid or partial isolation of human IgM from serum
Complete kit – IgM Purification Kit includes a pre-packed MBP column and ready-to-use buffers for convenient purification of at least 1.5mg IgM from 0.5mL ascites samples per column-use
Mild conditions – nondenaturing, near-neutral buffer conditions ensure recovery of functional antibodies
Reusable – stable ligand and resins can be regenerated and reused at least 10 times without significant decline in binding capacity
Includes:
Kit contains a 5mL MBP Agarose Column, column preparation buffer, IgM binding and elution buffers, and a column extender

Specifications
Agarose | |
Agarose Resin, Affinity | |
Lectin | |
Pierce Mannan Binding Protein Agarose | |
10 mL |
Pierce™ | |
Bottle | |
IgM Antibody | |
Upon receipt store product at 4°C. Product is shipped at ambient temperature. |
Suggestions de produits
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
For Research Use Only. Not for use in diagnostic procedures.