missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Immobilized Ficin (Agarose Resin)

Enable controlled antibody fragmentation, especially of mouse IgG1, with this sulfhydryl-specific ficin protease immobilized onto beaded agarose resin.

Brand:  Thermo Scientific™ 44881


Product Code. 10006813

  • £273.00 / 5mL
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Immobilized Ficin consists of a sulfhydryl-specific protease (ficin) that has been immobilized onto beaded agarose resin to enable controlled antibody fragmentation, especially of mouse IgG1.

Ficin, which is similar to papain, is a nonspecific, sulfhydryl protease isolated from fig latex. The molecular weight of Ficin is approximately 25,000 and it is effective at pH 4-9.5 with an optimum pH of 6.5. When IgG molecules are incubated with ficin in the presence of cysteine, one or more peptide bonds in the hinge region are split, producing either Fab fragments or F(ab')2 fragments. Mouse IgG1 is notably difficult to digest into functional Fab or F(ab')2 fragments using papain or pepsin, however, yields and immunoreactivity are better when digesting mouse IgG1 with ficin. Immobilized Ficin was specifically designed for cleavage of mouse IgG1 into or Fab or F(ab')2 fragments, and is advantageous because it virtually eliminates autolysis, eliminates contamination of a sample with the protease and allows control of the digestion by removing the ficin or controlling the flow of sample over an immobilized ficin column.

Highlights:

  • Can generate both Fab and F(ab')2 fragments from mouse IgG1
  • Reaction can be easily controlled
  • Antibody fragments are free of autodigestion products
  • Ficin contamination into antibody fragments is eliminated
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Content: 5 mL of settled resin.

Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C.
Liquid Suspension
5 mL
Ficin
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Immobilized Ficin, 5 Ml.

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Sodium chloride, Ficin

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.