missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Immobilized Pepsin (Agarose Resin)

Generate and purify Fab and F(ab')2 fragments from antibodies with acidic endopeptidase (pepsin) immobilized onto beaded agarose resin.

Brand:  Thermo Scientific™ 20343


Product Code. 10599103

  • £247.00 / 5mL
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Immobilized Pepsin consists of an acidic endopeptidase (pepsin) that has been immobilized onto beaded agarose resin to enable generation and purification of Fab and F(ab')2 fragments from antibodies.

Immobilized pepsin can be substituted for free pepsin in any application, and is advantageous because it virtually eliminates autolysis, eliminates contamination of a sample with the protease and allows control of the digestion by removing the pepsin. Immobilized pepsin is also more stable against heat-induced denaturation, resulting in longer maintenance of activity. The F(ab')2 Preparation Kit contains immobilized pepsin, other necessary reagents and an optimized protocol for human, rabbit and mouse antibody digestion.

Highlights:

  • Pepsin can cleave Fc fragments from IgG, resulting in F(ab')2 fragments
  • Broad specificity, including esterase activity
  • Easy to use - simply remove the agarose beads to stop the enzymatic digestion

Includes:

Kit contains immobilized pepsin, other necessary reagents and an optimized protocol for human, rabbit and mouse antibody digestion.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Upon receipt store product at 4-8°C. Product is shipped at ambient temperature.
Liquid Suspension
5 mL
Pepsin
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • IMMOBILIZED PEPSIN, 5 ML

Signal Word
  • Danger

Hazard Category
  • Respiratory sensitiser Category 1

Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

Supplemental information
  • MIXTURE LIST-Contains : Pepsin A

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.