Learn More
Thermo Scientific™ Pierce™ Kinase Enrichment Kit with ATP Probe
Desthiobiotin-ATP and -ADP probes to label, affinity purify and measure the active site of ATPases (kinases), including chaperones and metabolic enzymes.
Brand:  Thermo Scientific™ 88310
Product Code. 11870124
  • £788.00 / Each
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ Pierce Kinase Enrichment Kits utilize ActivX™ ATP or ADP Probes to covalently label the active site of ATPases, including chaperones and metabolic enzymes, to enable their selective enrichment using a desthiobiotin tag.
These products, based on ActivX ATP and ADP Probes, feature an amine-reactive nucleotide analog and a desthiobiotin (biotin analog) tag that facilitates selective labeling of lysines in the kinase active site and then subsequent enrichment and recovery of labeled protein. These features allow identification and profiling of target enzyme classes across samples or assessment of the specificity and affinity of enzyme inhibitors.
Highlights:
  • Specific – label only the conserved active-site lysines of nucleotide-binding proteins
  • Flexible – use for in vitro labeling of ATPase enzymes derived from cells or tissues
  • Compatible – use with Western blot or mass spectrometry (MS) workflows
Recommended for:
Profile small-molecule binding affinities and active-site inhibition in a dose-dependent manner; Identify dozens to hundreds of inhibitor targets and off-targets from tissues, cells and subcellular proteomes; Enrichment of enzymes based on function.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Multi-component kit with Desthiobiotin-ATP Probe
Protein Quantitation Assay
Kinase Substrate
16 reactions
Kinases (Multiple), Phosphodiesterases, ATPases, GTPases, Phosphatases
Biotin & Other Hapten
ActivX Desthiobiotin-ATP Probe is shipped separately with dry ice and stored at -80°C upon receipt. All other kit components are shipped at ambient temperature and stored at 4°C upon receipt.
Chemiluminescence, Mass Spectrometry, Western Blot
ActivX™, Pierce™
Pull-Down
Kinase Enrichment Kit
Tube, Spin Column